Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47750

Supelco

4-Amino-2,6-dinitrotoluene solution

certified reference material, 1000 μg/mL in acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813

grade

certified reference material
TraceCERT®

Quality Level

agency

EPA 8330

product line

TraceCERT®

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in acetonitrile

application(s)

environmental

format

single component solution

storage temp.

2-8°C

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service