Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47741-U

Supelco

ASTM® D5580 Quantitative Cal Mix 2

with internal standard, (Each component at nominal wt.% indicated.), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

agency

ASTM® D5580

packaging

ampule of 1 mL

concentration

(Each component at nominal wt.% indicated.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

room temp

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Benzene .45 % (w/w)
    Ethylbenzene .9 % (w/w)
    2-Hexanone 10 % (w/w)
    Toluene 9 % (w/w)
    1,2,4-Trimethylbenzene 9 % (w/w)
    2,2,4-Trimethylpentane 68.4 % (w/w)
    o-Xylene 2.25 % (w/w)

signalword

Danger

flash_point_f

10.0 °F

flash_point_c

-12.2 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service