Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47734-U

Supelco

ASTM® D5580 Quantitative Calibration Kit

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

agency

ASTM® D5580

CofA

current certificate can be downloaded

packaging

pkg of 1 kit

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

General description

ASTM® D5580 Quantitative Calibration Kit (ASTM Petroleum Standards) includes calibration blends provided with chromatogram and data verifying the purity and identity of the raw materials. All raw materials used are fully characterized, as described for ASTM D4815.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Each of the following components at the weight percent indicated.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Kit Components Only

Product No.
Description

  • Quantitative Calibration Mix 1 (47740-U) 1 mL

  • Quantitative Calibration Mix 2 (47741-U) 1 mL

  • Quantitative Calibration Mix 3 (47442-U) 1 mL

  • Quantitative Calibration Mix 4 (47743-U) 1 mL

  • Quantitative Calibration Mix 5 (47744-U) 1 mL

  • Valve Timing Calibration Mix (47731-U) 1 mL

  • Selectivity Check Standard (47732-U) 1 mL

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Ozone 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT RE 2 Inhalation - STOT SE 1 - STOT SE 3

target_organs

Central nervous system, Eyes,Central nervous system

Storage Class

3 - Flammable liquids

flash_point_f

10.0 °F - closed cup

flash_point_c

-12.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service