Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47727-U

Supelco

EPA 505/525 Update Pesticides Mix A

certified reference material, 500 μg/mL each component in acetone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

agency

EPA 505,525

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

500 μg/mL each component in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-30°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Alachlor
    Aldrin
    Lindane
    Dieldrin
    cis-Nonachlor
    trans-Nonachlor
    Simazine

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

1.4 °F - closed cup

flash_point_c

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service