Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47645

Supelco

2-Bromopropionic acid solution

certified reference material, 1000 μg/mL in methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
23151816

grade

certified reference material
TraceCERT®

agency

EPA 552.2

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type surrogate

packaging

ampule of 1 mL

concentration

1000 μg/mL in methyl tert-butyl ether

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C3H5BrO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)

InChI key

MONMFXREYOKQTI-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-27.4 °F - closed cup

flash_point_c

-33.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hari M R Gardimalla et al.
Chemical communications (Cambridge, England), (35)(35), 4432-4434 (2005-09-02)
Candida rugosa lipase immobilized on maghemite nanoparticles demonstrated high stereoselectivity in kinetic resolution of racemic carboxylates and improved long-term stability over its parent free enzyme, allowing the supported enzyme to be repeatedly used for a series of chiral resolution reactions.
J E McGrath et al.
Applied and environmental microbiology, 63(8), 3333-3335 (1997-08-01)
Type strains of the purple nonsulfur species Rhodospirillum rubrum, Rhodospirillum photometricum, and Rhodopseudomonas palustris grew phototrophically on a number of two- and three-carbon halocarboxylic acids in the presence of CO2, by reductive dehalogenation and assimilation of the resulting acid. Strains
E A Lock et al.
Archives of toxicology, 74(9), 547-554 (2000-12-29)
Oral administration of L-2-chloropropionic acid (L-CPA) to rats either as a single dose (750 mg/kg) or daily doses (250 mg/kg per day for 3 days) produces selective necrosis to the granule cell layer of the cerebellum. As part of a
O Brandsnes et al.
European journal of biochemistry, 126(2), 247-252 (1982-08-01)
The effect of temperature on the inactivation of liver alcohol dehydrogenase and on the alkylation of a model thiol free in solution by bromoacetate, 2-bromopropionate, 3-bromopropionate and 2-bromo-3-(5-imidazolyl)-propionate has been studied and the thermodynamic activation parameters calculated. All the bromoacids
Use of structure-activity relationships for probing biochemical mechanisms: glutathione transferase zeta conjugation of haloacids.
P D Swartz et al.
Advances in experimental medicine and biology, 500, 23-31 (2002-01-05)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service