Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47598-U

Supelco

EPA 552 Methyl Esters Mix

certified reference material, 1000 μg/mL each component in methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

agency

EPA 552

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in methyl tert-butyl ether

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA 552 Methyl Esters Mix is a multi-component solution (1000 μg/mL each component in methyl tert-butyl ether) containing methyl bromoacetate, methyl bromochloroacetate, methyl chloroacetate, methyl dibromoacetate, methyl dichloroacetate, and methyl trichloroacetate as certified reference materials (CRMs). EPA 552 Methyl Esters Mix is typically used to quantitatively calibrate capillary gas chromatographs used to determine the byproducts of drinking water disinfection. These byproducts have the potential to cause adverse human health effects.
The US EPA method 552 is one of the methods used to monitor the levels of disinfection byproducts. Laboratories that support water systems needing to determine disinfection byproducts are required to use this method.
US EPA 500 Series methods are analytical methods for identifying and quantifying organic contaminants such as volatile organic compounds (VOCs), pesticides, synthetic organic compounds (SOCs), and trihalomethane disinfection byproducts (THMs) in raw source water, finished drinking water, and drinking water at all stages of treatment.

Application

EPA 552 Methyl Esters Mix is used as a CRM to quantify disinfection by-products (DBPs) such as haloacetic acids in drinking water by gas chromatography-mass spectrometry (GC-MS) technique.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Methyl bromoacetate
    Methyl bromochloroacetate
    Methyl chloroacetate
    Methyl dibromoacetate
    Methyl dichloroacetate
    Methyl trichloroacetate

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-18.4 °F - closed cup

flash_point_c

-28 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quenching of chlorination disinfection by-product formation in drinking water by hydrogen peroxide
Batterman S, et al.
Water Research, 34(5), 1652-1658 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service