Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47557-U

Supelco

Chlorinated Pesticides Mix

in isooctane (varied), TraceCERT®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

in isooctane (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

multi-component solution

Storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Chlorinated Pesticides Mix is suitable for use in environmental and food residue analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Aldrin 50 μg/mL
    α-1,2,3,4,5,6-Hexachlorocyclohexane
    β-BHC
    Lindane 25 μg/mL
    1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane 200 μg/mL
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene 100 μg/mL
    2,4′-DDT 225 μg/mL
    4,4′-DDT 260 μg/mL
    Dieldrin 120 μg/mL
    Endrin 200 μg/mL
    Heptachlor 25 μg/mL
    Heptachlor exo-epoxide 80 μg/mL
    See All (13)

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

10.4 °F - closed cup

flash_point_c

-12 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service