Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47538

USP 467 OVI Mix

high concentration

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
Pricing and availability is not currently available.

feature

standard type 5 component

packaging

ampule of 1 mL

concentration

in DMSO

storage temp.

2-8°C

Analyte

Description

    Benzene 1000 μg/mL
    Chloroform 500 μg/mL
    Dichloromethane 5000 μg/mL
    1,4-Dioxane 1000 μg/mL
    Trichloroethylene 1000 μg/mL

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service