Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47375

Supelco

EPA 8150 Herbicide Methyl Derivatives Mix

certified reference material, in hexane (varied conc.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813

grade

certified reference material
TraceCERT®

Quality Level

agency

EPA 8150,8151,8151A

product line

TraceCERT®

feature

standard type calibration

packaging

ampule of 1 mL

concentration

in hexane (varied conc.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    4-Chloro-2-methylphenoxyacetic acid 200 μg/mL
    2,4-Dichlorophenoxyacetic acid 200 μg/mL
    4-(2,4-Dichlorophenoxy)butyric acid 200 μg/mL
    2,2-Dichloropropionic acid 200 μg/mL
    Dicamba 200 μg/mL
    Dichlorprop 200 μg/mL
    Dinoseb 200 μg/mL
    Mecoprop 2000 μg/mL
    2,4,5-Trichlorophenoxyacetic acid 200 μg/mL
    2-(2,4,5-Trichlorophenoxy)propionic acid 200 μg/mL
    See All (10)

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-14.8 °F - closed cup

flash_point_c

-26.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service