47335-U
Chloral hydrate solution
1000 μg/mL in acetonitrile, analytical standard
About This Item
Recommended Products
grade
analytical standard
agency
EPA 551,8260
CofA
current certificate can be downloaded
feature
standard type calibration
packaging
ampule of 1 mL
concentration
1000 μg/mL in acetonitrile
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
storage temp.
2-30°C
SMILES string
OC(O)C(Cl)(Cl)Cl
InChI
1S/C2H3Cl3O2/c3-2(4,5)1(6)7/h1,6-7H
InChI key
RNFNDJAIBTYOQL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Chloral hydrate solution (Method: EPA 8260) is utilized for identifying and quantifying volatile organic compounds on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites. The pre-requisite of EPA 8260 method is the use of gas chromatography with a selective detector or gas chromatography/mass spectrometry.
Application
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service