Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47327

Supelco

ASTM®/EPA Aromatics Internal Standard Mix

(each component at wt% indicated), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813

grade

analytical standard

agency

ASTM® D5769

CofA

current certificate can be downloaded

packaging

pkg of 5.0 mL

concentration

(each component at wt% indicated)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
petroleum

format

multi-component solution

storage temp.

2-30°C

General description

ASTM®/EPA Aromatics Internal Standard Mix may be used for the determination of aromatics in Gasoline by GC-MS.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Benzene-D6 40 % (w/w)
    Ethylbenzene-d10 40 % (w/w)
    Naphthalene-d8 20 % (w/w)

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1 - STOT RE 2

target_organs

Blood, hearing organs

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service