Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47116

Supelco

Menhaden fish oil

analytical standard

Synonym(s):

Fish oil from menhaden, Menhaden oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164500

biological source

fish oil (menhaden)

grade

analytical standard

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.93 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

This reference material is offered as a qualitative standard. Use the chromatographic fingerprint of our reference material to check for the presence of Menhaden fish oil in your sample. A Certificate of Composition which includes a chromatogram is shipped with each product.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

12 - Non Combustible Liquids

wgk_germany

awg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pedro Beca-Carretero et al.
Marine pollution bulletin, 134, 27-37 (2018-01-15)
Global warming is emerging as one of the most critical threats to terrestrial and marine species worldwide. This study assessed the effects of simulated warming events in culture on two seagrass species, Posidonia oceanica and Cymodocea nodosa, which play a
Snehal Gite et al.
Nutritional neuroscience, 22(8), 551-568 (2018-01-31)
Objectives: To search for novel compounds that will protect neuronal cells under stressed conditions that may help to restore neuronal plasticity. Methods: A model of corticosterone (CORT)-induced stress in human neuroblastoma cells (SH-SY5Y) was used to compare the efficacy of
Sang Lee et al.
Journal of pediatric surgery, 43(11), 2010-2015 (2008-10-31)
Bile duct obstruction and subsequent cholestasis produces hepatocellular injury and an inflammatory response. Fatty acid constitution of cell membranes plays a major role in the inflammatory cascade. Omega-3 fatty acids are antiinflammatory. We proposed that omega-3 fatty acid supplementation would
Paola Dugo et al.
Journal of chromatography. A, 1259, 227-236 (2012-04-17)
A non-aqueous reversed phase high performance liquid chromatography method was developed, and optimized for triacylglycerol analysis in a Brevoortia tyrannus (menhaden) oil sample. Four columns were serially coupled to tackle such a task, for a total length of 60 cm
Constance Lay Lay Saw et al.
Asian Pacific journal of cancer prevention : APJCP, 12(12), 3331-3334 (2011-01-01)
Numerous epidemiological studies suggest that frequent consumption of fish would decrease certain major inflammatory-related chronic diseases including cancer. To investigate the cancer chemoprotective effect of fish oil (FO) in Transgenic Adenocarcinoma of Mouse Prostate (TRAMP) mice fed a FO diet

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service