Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47115-U

Supelco

Lard oil

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

oleic acid

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Characterized lard oil samples are available for use as controls or check samples for fatty acid methyl ester (FAME) analyses. These reference materials can be used as qualitative standards. They provide an excellent means of standardizing the lipid procedures and comparing the results to others. Packed in an amber ampule under nitrogen.

Application

Lard oil may be used as an analytical reference standard for the determination of the analyte in milk samples by high performance liquid chromatography technique.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

12 - Non Combustible Liquids

wgk_germany

awg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of human milk triacylglycerols by high-performance liquid chromatography with light-scattering detection.
Pons SM, et al.
Journal of Chromatography A, 823(1-2), 475-482 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service