46938-U
Malonic acid
analytical standard
Synonym(s):
Propanedioic acid
About This Item
Recommended Products
grade
analytical standard
assay
99.50%
CofA
current certificate can be downloaded
packaging
ampule of 500 mg
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
mp
132-135 °C (dec.) (lit.)
density
1.619 g/cm3 at 25 °C
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
storage temp.
2-30°C
SMILES string
OC(=O)CC(O)=O
InChI
1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)
InChI key
OFOBLEOULBTSOW-UHFFFAOYSA-N
Gene Information
human ... SRC(6714)
Looking for similar products? Visit Product Comparison Guide
General description
Application
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
314.6 °F - closed cup
flash_point_c
157 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service