Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46862-U

Supelco

Pesticide Mix A-2

certified reference material, in hexane: toluene (99:1) (varied)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

ampule of 1 mL

concentration

in hexane: toluene (99:1) (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    α-BHC
    Gamma-BHC
    4,4′-DDT 10 μg/mL
    Decachlorobiphenyl 10 μg/mL
    Dieldrin 10 μg/mL
    α-Endosulfan 5 μg/mL
    Endrin 10 μg/mL
    Heptachlor 5 μg/mL
    Lindane 5 μg/mL
    Methoxychlor 50 μg/mL
    2,4,5,6-Tetrachloro-m-xylene 5 μg/mL
    4,4′-DDD
    See All (12)

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT RE 2 - STOT SE 3

target_organs

Central nervous system, Endocrine system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-14.8 °F - closed cup

flash_point_c

-26 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service