Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46861-U

Supelco

Acid Herbicide Mix

in methanol (varied), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

in methanol (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

General description

Acid Herbicide Mix (Method: EPA 8150/8151/8151A) is utilized for identifying and quantifying chlorinated herbicides on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites. The pre-requisite of EPA 8150/8151/8151A method is the use of gas chromatography with a selective detector or gas chromatography/mass spectrometry.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    4-Amino-3,5,6-trichloropyridine-2-carboxylic acid 100 μg/mL
    4-Chloro-2-methylphenoxyacetic acid 10000 μg/mL
    Dicamba 10 μg/mL
    2,4-Dichlorophenoxyacetic acid 100 μg/mL
    4-(2,4-Dichlorophenoxy)butyric acid 100 μg/mL
    2-(2,4-Dichlorophenoxy)propionic acid 100 μg/mL
    2,2-Dichloropropionic acid 250 μg/mL
    Dinoseb 50 μg/mL
    Mecoprop 10000 μg/mL
    Pentachlorophenol 50 μg/mL
    2-(2,4,5-Trichlorophenoxy)propionic acid 200 μg/mL
    2,4,5-Trichlorophenoxyacetic acid 200 μg/mL
    See All (12)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

51.8 °F

flash_point_c

11 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service