Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44803

Supelco

PCB kit - high conc.

1000 μg/mL in isooctane (each solution), analytical standard

Synonym(s):

PCB Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

analytical standard

agency

EPA 8082
MISA (Canada) Group 27

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

pkg of 1 kit

concentration

1000 μg/mL in isooctane (each solution)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

Storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Kit Components Also Available Separately

Product No.
Description
SDS

  • 44805Aroclor 1232 solution, certified reference material, 1000 μg/mL in isooctane 1 mLSDS

  • 44806Aroclor 1242 solution, certified reference material, 1000 μg/mL in isooctane 1 mLSDS

  • 44807Aroclor 1248 solution, certified reference material, 1000 μg/mL in isooctane 1 mLSDS

  • 44808Aroclor 1254 solution, certified reference material, 1000 μg/mL in isooctane 1 mLSDS

  • 44809Aroclor 1260 solution, certified reference material, 1000 μg/mL in isooctane 1 mLSDS

  • 44810Aroclor 1262 solution, certified reference material, 1000 μg/mL in isooctane 1 mLSDS

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

flash_point_f

17.6 °F

flash_point_c

-8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service