Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44588

Supelco

Naphthenes Mix

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

analytical standard

agency

ASTM® D6729,D6730,D6733

packaging

pkg of 0.1 mL

concentration

(typical values shown)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
petroleum

format

multi-component solution

storage temp.

2-8°C

General description

This is a 30 component mix of known naphthenes. It is intended for use in identifying naphthenes in complex hydrocarbon mixtures.

  • Formulation is weight percent, and will vary lot to lot.
  • A detailed data sheet listing components by weight percent, mole percent, and liquid volume percent, retention times and other formulaiton is provided with the product.
  • Supplied in a crimp-top vial with hole cap and septa.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    n-Butylcyclopentane 3.724 wt. %
    Cyclohexane 5.4 wt. %
    Cyclopentane 4.97 wt. %
    cis-1,2-Dimethylcyclohexane 3.725 wt. %
    trans-1,2-Dimethylcyclohexane 1.669 wt. %
    trans-1,4-Dimethylcyclohexane 3.557 wt. %
    1,1-Dimethylcyclopentane 3.483 wt. %
    trans-1,2-Dimethylcyclopentane 1.468 wt. %
    cis-1,3-Dimethylcyclopentane .604 wt. %
    trans-1,3-Dimethylcyclopentane 2.749 wt. %
    Ethylcyclopentane 3.575 wt. %
    1-Ethyl-1-methylcyclopentane 1.076 wt. %
    Isobutylcyclohexane 5.639 wt. %
    Isobutylcyclopentane 3.727 wt. %
    Isopropylcyclohexane 5.77 wt. %
    Isopropylcyclopentane 3.511 wt. %
    Methylcyclohexane 5.711 wt. %
    Methylcyclopentane 3.269 wt. %
    t-1-Methyl-2-(4MP)cyclopentane 3.795 wt. %
    trans-1-Methyl-2-propylcyclohexane 3.289 wt. %
    Propylcyclopentane 3.677 wt. %
    1,1,2-Trimethylcyclohexane 3.371 wt. %
    1,1,4-Trimethylcyclohexane 3.68 wt. %
    cis,trans,cis-1,2,4-Trimethylcyclohexane 3.541 wt. %
    cis,trans,trans-1,2,4-Trimethylcyclohexane 3.634 wt. %
    cis,cis,cis-1,3,5-Trimethylcyclohexane 3.558 wt. %
    cis,cis,cis-1,2,3-Trimethylcyclopentane .808 wt. %
    cis,trans,cis-1,2,3-Trimethylcyclopentane 1.586 wt. %
    cis,trans,cis-1,2,4-Trimethylcyclopentane 1.673 wt. %
    cis,trans,trans-1,2,4-Trimethylcyclopentane 3.76 wt. %
    See All (30)

analytical column

standard

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-4.0 °F - closed cup

flash_point_c

-20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service