Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44587

Supelco

Aromatics Mix

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

analytical standard

agency

ASTM® D6729,D6730,D6733

packaging

pkg of 0.1 mL

concentration

(typical values shown)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
petroleum

format

multi-component solution

storage temp.

2-8°C

General description

This is a 37 component mix of known aromatics. It is intended for use in identifying aromatics in complex hydrocarbon mixtures.

  • Formulation is weight percent, and will vary lot to lot.
  • A detailed data sheet listing components by weight percent, mole percent, and liquid volume percent, retention times and other formulaiton is provided with the product.
  • Supplied in a crimp-top vial with hole cap and septa.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Benzene 7.099 wt. %
    Butylbenzene 2.196 wt. %
    sec-Butylbenzene 2.238 wt. %
    tert-Butylbenzene 4.57 wt. %
    1-tert-Butyl-3,5-dimethylbenzene 2.15 wt. %
    1-tert-Butyl-4-ethylbenzene 2.225 wt. %
    tert-1-Butyl-2-methylbenzene .764 wt. %
    1,2-Diethylbenzene 1.088 wt. %
    1,2-Dimethyl-3-ethylbenzene 2.176 wt. %
    1,2-Dimethyl-4-ethylbenzene 2.243 wt. %
    1,3-Dimethyl-2-ethylbenzene 1.138 wt. %
    1,3-Dimethyl-5-ethylbenzene 2.186 wt. %
    1,4-Dimethyl-2-ethylbenzene 2.272 wt. %
    Ethylbenzene 6.723 wt. %
    Hexylbenzene 4.463 wt. %
    Isobutylbenzene 4.39 wt. %
    Cumene 2.211 wt. %
    2-Methylbutylbenzene 1.137 wt. %
    1-Methyl-2-ethylbenzene 2.237 wt. %
    1-Methyl-3-ethylbenzene 2.188 wt. %
    1-Methyl-4-ethylbenzene 2.156 wt. %
    1-Methyl-2-isopropylbenzene 1.12 wt. %
    1-Methyl-3-isopropylbenzene 1.102 wt. %
    1-Methyl-4-isopropylbenzene 1.065 wt. %
    1-Methyl-2-n-propylbenzene 2.231 wt. %
    1-Methyl-3-n-propylbenzene 2.1 wt. %
    1-Methyl-4-n-propylbenzene 2.212 wt. %
    Pentylbenzene 4.443 wt. %
    Propylbenzene 4.532 wt. %
    1,2,4,5-Tetramethylbenzene .238 wt. %
    Toluene 4.55 wt. %
    1,2,4-Triethylbenzene 1.107 wt. %
    1,3,5-Triethylbenzene 4.525 wt. %
    1,2,4-Trimethylbenzene 2.523 wt. %
    1,3,5-Trimethylbenzene 1.12 wt. %
    o-Xylene 2.245 wt. %
    m-Xylene 2.256 wt. %
    p-Xylene 4.784 wt. %
    See All (38)

analytical column

standard

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1A - Eye Dam. 1 - Flam. Liq. 3 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

target_organs

Blood, Respiratory system

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service