Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442843

Supelco

Fluoranthene-d10

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D10
CAS Number:
Molecular Weight:
212.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

384 °C (lit.)

mp

110-113 °C (lit.)

application(s)

environmental

format

neat

mass shift

M+10

storage temp.

2-30°C

SMILES string

[2H]c1c([2H])c([2H])c-2c(c1[2H])-c3c([2H])c([2H])c([2H])c4c([2H])c([2H])c([2H])c-2c34

InChI

1S/C16H10/c1-2-8-13-12(7-1)14-9-3-5-11-6-4-10-15(13)16(11)14/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

GVEPBJHOBDJJJI-LHNTUAQVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

388.4 °F - closed cup

flash_point_c

198.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service