Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442809

Supelco

Toluene-d8

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CD3
CAS Number:
Molecular Weight:
100.19
Beilstein/REAXYS Number:
1909300
EC Number:
MDL number:
UNSPSC Code:
41115716
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.494 (lit.)

bp

110 °C (lit.)

density

0.943 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

mass shift

M+8

storage temp.

2-30°C

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])[2H]

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3/i1D3,2D,3D,4D,5D,6D

InChI key

YXFVVABEGXRONW-JGUCLWPXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F - closed cup

flash_point_c

4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyong Lu et al.
Molecules (Basel, Switzerland), 19(9), 14292-14303 (2014-09-13)
Gated molecular encapsulations, with baskets of type 1, are postulated to occur by the mechanism in which solvent molecule penetrates the inner space of 1, through one of its apertures, while the residing guest simultaneously departs the cavity. In the
Valerie L Colley et al.
Drug testing and analysis, 7(3), 241-246 (2014-10-11)
South American 'crack' cocaine, produced directly from coca leaf, can be distinguished from US domestically produced crack on the basis of occluded solvent profiles. In addition, analysis of domestically produced crack indicates the solvents that were used for cocaine hydrochloride
Tiina J Kauppila et al.
Journal of the American Society for Mass Spectrometry, 25(11), 1870-1881 (2014-09-25)
A novel, gas-tight API interface for gas chromatography-mass spectrometry was used to study the ionization mechanism in direct and dopant-assisted atmospheric pressure photoionization (APPI) and atmospheric pressure laser ionization (APLI). Eight analytes (ethylbenzene, bromobenzene, naphthalene, anthracene, benzaldehyde, pyridine, quinolone, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service