Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442725

Supelco

Octachloronaphthalene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352200

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 20 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

InChI

1S/C10Cl8/c11-3-1-2(5(13)9(17)7(3)15)6(14)10(18)8(16)4(1)12

InChI key

RTNLUFLDZOAXIC-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Orlikowska et al.
Chemosphere, 75(9), 1196-1205 (2009-03-17)
The amounts, profiles and origin of CNs (from triCNs to octaCN) sequestered in Scots pine needles collected from 25 spatially distant sites in Poland have been studied based on congener-specific data obtained after a several clean-up and fractionation steps and
Photolysis of octachloronaphthalene in hexane.
Y S Keum et al.
Bulletin of environmental contamination and toxicology, 72(5), 999-1005 (2004-07-23)
R F Lopshire et al.
Toxicology and industrial health, 12(3-4), 375-391 (1996-05-01)
The exchange reaction of chlorine by oxygen with polychlorinated biphenyl (PCB) anions was explored as a method for composition-selective detection of PCB congeners in a gas chromatograph/tandem mass spectrometer (GC/MS/MS) experiment. The odd-electron molecular anions of PCBs react with molecular
M A Campbell et al.
Toxicology, 22(2), 123-132 (1981-01-01)
Administration of octachloronaphthalene to immature male Wistar rats resulted in a dose-dependent increase in several enzymic, electrophoretic and spectral parameters associated with induction of the hepatic microsomal enzymes. Compared to corn-oil (control)treated animals octachloronaphthalene (150 mumol . kg-1) induced hepatic
A Belfroid et al.
Ecotoxicology and environmental safety, 31(3), 185-191 (1995-08-01)
The uptake and elimination of three superlipophilic compounds (hexabromobenzene, PCB153, and octachloronaphthalene) after dietary uptake was studied in earthworms (Eisenia andrei). All three compounds were taken up from the food, although they did not significantly accumulate despite their hydrophobicity. Both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service