Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442708

Supelco

Tetradecane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12CH3
CAS Number:
Molecular Weight:
198.39
Beilstein/REAXYS Number:
1733859
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

vapor density

6.83 (vs air)

vapor pressure

1 mmHg ( 76.4 °C)

CofA

current certificate can be downloaded

autoignition temp.

455 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.429 (lit.)

bp

252-254 °C (lit.)

mp

5.5 °C (lit.)

density

0.762 g/mL at 20 °C (lit.)

application(s)

environmental
petroleum

format

neat

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCC

InChI

1S/C14H30/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3-14H2,1-2H3

InChI key

BGHCVCJVXZWKCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tetradecane may be used as an internal standard in investigating the geographical discrimination and commercial categorization of saffron using high-performance liquid chromatography (HPLC), near-infrared spectroscopy (NIRS) and gas chromatography coupled with mass spectrometry (GC-MS).[1]

Other Notes

Product 442708 has been discontinued. Product 87139 may be an acceptable replacement.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

supp_hazards

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

215.6 °F - closed cup

flash_point_c

102 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An integrated approach combining HPLC, GC/MS, NIRS, and chemometrics for the geographical discrimination and commercial categorization of saffron.
Liu J, et al.
Food Chemistry, 253, 284-292 (2018)
John J Beck et al.
Journal of agricultural and food chemistry, 57(9), 3749-3753 (2009-04-07)
Nonpareil almonds, Prunus dulcis , account for the largest percentage of almond varieties grown in the Central Valley of California. Several studies have investigated the various nonvolatile and volatile components of various plant parts; however, the volatile organic compound (VOC)
Yong Zhou et al.
Journal of the American Chemical Society, 131(7), 2456-2457 (2009-02-10)
In this Communication, we report one-pot, high yield formation of unprecedented 1-D necklace-like chains consisting of hybrid Pd nanocrystals (NCs) embedded in spheres of a peptidic lipid. We employed the synthetic peptidic lipid, 2-(2-tetradecanamidoacetamido)acetic acid (1). We found that in
Jacques Joubert et al.
Bioorganic & medicinal chemistry, 16(19), 8952-8958 (2008-09-23)
In recent years polycyclic compounds have been shown to exhibit pharmacological profiles of importance in the symptomatic and proposed curative treatment of neurodegenerative diseases (e.g., Parkinson's and Alzheimer's disease). These structures also show modification and improvement of the pharmacokinetic and
Vishwakarma Singh et al.
The Journal of organic chemistry, 74(16), 6092-6104 (2009-07-21)
A general and stereoselective methodology for the synthesis of bridged bicyclic octenones having various types of alkenyl chains and a tricyclic framework of secoatisanes and higher analogues is reported. In situ generation and cycloaddition of 2-allyl-6,6-spiroepoxycyclohexadienones with ethyl acrylate gave

Protocols

Separation of Decane; Dodecane; Tetradecane; Hexadecane; Octadecane; Eicosane; Docosane; Tetracosane; Hexacosane; Octacosane

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service