Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442694

Supelco

Nonane

analytical standard

Synonym(s):

n-Nonane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH3
CAS Number:
Molecular Weight:
128.26
Beilstein/REAXYS Number:
1696917
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

vapor density

4.41 (vs air)

vapor pressure

0.18 psi ( 37.7 °C)

CofA

current certificate can be downloaded

autoignition temp.

401 °F

expl. lim.

2.9 %

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.405 (lit.)

bp

151 °C (lit.)

mp

−53 °C (lit.)

density

0.718 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

storage temp.

room temp

SMILES string

CCCCCCCCC

InChI

1S/C9H20/c1-3-5-7-9-8-6-4-2/h3-9H2,1-2H3

InChI key

BKIMMITUMNQMOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Product 442694 has been discontinued. Product 74250 may be an acceptable replacement.

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

87.8 °F - closed cup

flash_point_c

31.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Franck Slowinski et al.
Organic letters, 12(21), 5004-5007 (2010-10-14)
New azabicyclo[2.2.1]heptane and -[3.3.1]nonane derivatives containing a pyridinyl substituent at the bridgehead position have been synthesized via an efficient ten chemical steps pathway. Both chemical series were then evaluated in vitro for their affinity at α7 nicotinic receptors revealing nanomolar
Lip Kim Lee et al.
Applied and environmental microbiology, 76(3), 794-802 (2009-12-17)
Polybrominated diphenyl ethers (PBDEs) have attracted attention recently due to their proven adverse effects on animals and their increasing concentrations in various environmental media and biota. To gain insight into the fate of PBDEs, microcosms established with soils and sediments
Arif Baran et al.
The Journal of organic chemistry, 77(3), 1244-1250 (2012-01-11)
1,3,3a,7a-Tetrahydro-2-benzofuran was used as key compound for the synthesis of various bishomoinositol derivatives. The diene was subjected to an epoxidation reaction for further functionalization of the diene unit. The bisepoxide obtained was submitted to a ring-opening reaction with acid in
Arif Baran et al.
The Journal of organic chemistry, 77(11), 5086-5097 (2012-05-23)
Transformation of cyclohexa-2,4-diene-1,2-diylbis(methylene) diacetate to various carbasugars is described. Photooxygenation of a cyclohexadiene derivative gave a bicyclicendoperoxide, which was reduced with thiourea to [2-[(acetyloxy)methyl]cyclohexa-2,4-dien-1-yl]methyl acetate. Epoxidation of the remaining double bond followed by epoxide ring-opening and hydrolysis of the acetate
Cesar A López et al.
Carbohydrate research, 364, 1-7 (2012-11-07)
The molecular dynamics simulation technique was used to study the folding and complexation process of a short amylose fragment in the presence of lipids. In aqueous solution, the amylose chain remains as an extended left-handed helix. After the addition of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service