Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442373

Supelco

3,4,5-Trichlorophenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl3O
CAS Number:
Molecular Weight:
197.45
EC Number:
UNSPSC Code:
12352200

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 25 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

InChI

1S/C6H3Cl3O/c7-4-1-3(10)2-5(8)6(4)9/h1-2,10H

InChI key

GBNHEBQXJVDXSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4,5-Trichlorophenol is a needle shaped type of chlorinated phenol.[1]
3,4,5-Trichlorophenol is an environmental pollutant belonging to the class of chlorophenols. Chlorophenols are formed as metabolites of chlorinated phenoxy acid herbicides and also exist as byproducts from the chlorination of drinking water.[2] They find applications in the production of pharmaceuticals, dyes, polymers, etc.[3]

Application

3,4,5-Trichlorophenol may be used as an analytical reference standard for the quantification of the analyte in landfill leachates[4] and aqueous samples[2][3] using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Environmental Analysis (2001)
Manjia Chen et al.
Ecotoxicology and environmental safety, 205, 111328-111328 (2020-09-21)
Understanding the degradation of pentachlorophenol (PCP) by indigenous microorganisms stimulated by an electron donor and shuttle in paddy soil, and the influences of PCP/electron donor/shuttle on the native microbial community are important for biodegradation and ecological and environmental safety. Previous
Lukas Schmidt et al.
Analytica chimica acta, 965, 123-130 (2017-04-04)
The determination of chlorophenols in urine is an established approach for the assessment of human exposure to these ubiquitous environmental pollutants. However, an analytical procedure which enables the separate determination of all components of this substance group was still lacking.
Development and optimization of a method for the analysis of phenols and chlorophenols from aqueous samples by gas chromatography-mass spectrometry, after solid-phase extraction and trimethylsilylation.
Kovacs A, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 99(1), 125-131 (2011)
Determination of chlorophenols in water samples using simultaneous dispersive liquid--liquid microextraction and derivatization followed by gas chromatography-electron-capture detection.
Fattahi N, et al.
Journal of Chromatography A, 1157(1-2), 23-29 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service