Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442360

Supelco

2-Methylpentane

analytical standard

Synonym(s):

‘Isohexane’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
86.18
Beilstein/REAXYS Number:
1730735
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

3 (vs air)

vapor pressure

6.77 psi ( 37.7 °C)

autoignition temp.

583 °F

expl. lim.

7 %

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.371 (lit.)

bp

62 °C (lit.)

mp

−154 °C (lit.)

density

0.653 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

Storage temp.

room temp

SMILES string

CCCC(C)C

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

Inchi Key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

19.4 °F - closed cup

flash_point_c

-7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea N Giordano et al.
The journal of physical chemistry. A, 119(15), 3545-3555 (2015-03-27)
We report the solvent-dependent dynamics of carbonyl site exchange for Fe(CO)3(η(4)-norbornadiene) (FeNBD) in a series of linear and nonlinear alkanes. The barrier to exchange is very low (∼1.5 kcal/mol), and the resulting carbonyl dynamics are rapid enough to lead to
2-Methylpentane (isohexane). CAS# 107-83-5.
J B Galvin et al.
Journal of toxicology and environmental health. Part A, 58(1-2), 81-92 (1999-10-28)
M Imbriani
Giornale italiano di medicina del lavoro, 15(5-6), 133-138 (1993-09-01)
We have considered the opportunity of using some inert vapours (2-3 Dimethylbutane, 2 Methylpentane, n-Hexane) to measure the pulmonary capillary blood flow (Qc). The measurements were performed by means of a gas chromatograph connected with a mass spectrometer. The alpha
M Ojala et al.
Rapid communications in mass spectrometry : RCM, 14(11), 994-998 (2000-06-09)
A method using purge-and-membrane mass spectrometry (PAM-MS) was developed for the analysis of residual solvents in pharmaceutical products. The method combines dynamic headspace and membrane inlet mass spectrometry. The limits of detection for the compounds studied, benzene, toluene, chloroform, 2-pentene
Y C Hsu et al.
Chemosphere, 42(3), 227-234 (2000-12-02)
Motor vehicle emission factors of carbon monoxide (CO) and non-methane volatile organic compounds (NMVOCs) were calculated inside the Chung-Cheng Tunnel of Kaohsiung in Taiwan. The results were compared with those model predictions from the Mobile Taiwan 2.0 model. Individual concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service