All Photos(1)
About This Item
Linear Formula:
CH2=CH(CH2)17CH3
CAS Number:
Molecular Weight:
280.53
Beilstein/REAXYS Number:
1768357
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
grade
analytical standard
packaging
ampule of 1000 mg
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
bp
151 °C/1.5 mmHg (lit.)
mp
26-30 °C (lit.)
application(s)
environmental
petroleum
format
neat
storage temp.
room temp
SMILES string
CCCCCCCCCCCCCCCCCCC=C
InChI
1S/C20H40/c1-3-5-7-9-11-13-15-17-19-20-18-16-14-12-10-8-6-4-2/h3H,1,4-20H2,2H3
InChI key
VAMFXQBUQXONLZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
1-Eicosene has been used as internal standard in the determination and analysis of aliphatic hydrocarbons and linear alkylbenzenes in sewage sludge-amended soil using high performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Analysis and fate of aliphatic hydrocarbons, linear alkylbenzenes, polychlorinated biphenyls and polycyclic aromatic hydrocarbons in sewage sludge-amended soils
Mangas.E, et al.
Chemosphere, 36, 61-72 (1998)
Morten T Nielsen et al.
Applied and environmental microbiology, 80(23), 7258-7265 (2014-09-23)
Forskolin is a promising medicinal compound belonging to a plethora of specialized plant metabolites that constitute a rich source of bioactive high-value compounds. A major obstacle for exploitation of plant metabolites is that they often are produced in small amounts
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service