Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442261

Supelco

1-Chlorooctane

analytical standard

Synonym(s):

Octyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7Cl
CAS Number:
Molecular Weight:
148.67
Beilstein/REAXYS Number:
1697464
EC Number:
MDL number:
UNSPSC Code:
41121813
PubChem Substance ID:

grade

analytical standard

Quality Level

vapor pressure

1 mmHg ( 22.2 °C)

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.430 (lit.)

bp

183 °C (lit.)

mp

−61 °C (lit.)

density

0.875 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

CCCCCCCCCl

InChI

1S/C8H17Cl/c1-2-3-4-5-6-7-8-9/h2-8H2,1H3

InChI key

CNDHHGUSRIZDSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Health hazardEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

141.8 °F - closed cup

flash_point_c

61 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R King et al.
British journal of pharmacology, 157(6), 974-983 (2009-06-03)
Monoacylglycerol lipase (MGL) is a presynaptic serine hydrolase that inactivates the endocannabinoid neurotransmitter, 2-arachidonoyl-sn-glycerol. Recent studies suggest that cysteine residues proximal to the enzyme active site are important for MGL function. In the present study, we characterize the role of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service