Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442237

Supelco

1,3,5-Trinitrobenzene

analytical standard, wetted material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3N3O6
CAS Number:
Molecular Weight:
213.10
EC Number:
UNSPSC Code:
12000000

grade

analytical standard

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

room temp

InChI

1S/C6H3N3O6/c10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15/h1-3H

InChI key

UATJOMSPNYCXIX-UHFFFAOYSA-N

Application

1,3,5-Trinitrobenzene may be used as a standard:
  • For analyzing the toxic effects of TNT in sediments on two freshwater invertebrates, Chironomus tentans and Hyalella azteca. [1]
  • In the determination of 1,3,5-trinitrobenzene in environmental samples using high performance liquid chromatography coupled with atmospheric pressure ionization mass spectrometry (HPLC-API-MS).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Expl. 1.1 - STOT RE 2

Storage Class

1 - Explosive hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele Biagioli et al.
Nutrients, 12(7) (2020-07-08)
Dysbiosis is commonly detected in patients with inflammatory bowel disease (IBD), supporting the concept that a dysregulated immune reaction to bacterial antigens has a pathogenic role in the development of intestinal inflammation. In the present study, we have investigated the
Jakub Nawała et al.
The Science of the total environment, 708, 135198-135198 (2019-12-10)
After World War II, conventional and chemical ammunition containing mainly secondary and primary explosives was dumped in the sea. Explosives have medium toxicity to aquatic organisms, earthworms and indigenous soil microorganisms. Therefore, environmental monitoring is required, especially for dumped munitions.
Yiqun Chen et al.
Chemosphere, 236, 124365-124365 (2019-07-22)
Nitroaromatic compounds (NACs) are widely distributed in the environment and are considered toxic or carcinogenic. However, little attention has been paid to the binding interactions between NACs and biomacromolecules (e.g., proteins). Here we investigated the effects of three model NACs
Toxicity of the explosives 2, 4, 6?trinitrotoluene, hexahydro?1, 3, 5?trinitro?1, 3, 5?triazine, and octahydro?1, 3, 5, 7?tetranitro?1, 3, 5, 7?tetrazocine in sediments to Chironomus tentans and Hyalella azteca: Low?dose hormesis and high?dose mortality.
Steevens J A, et al.
Environmental Toxicology and Chemistry / Setac, 21(7), 1475-1482 (2002)
Application of spectral libraries for high-performance liquid chromatography?atmospheric pressure ionisation mass spectrometry to the analysis of pesticide and explosive residues in environmental samples.
Schreiber A, et al.
Journal of Chromatography A, 869(1-2), 411-425 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service