Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42893

Supelco

Tetracosanoic acid

analytical standard

Synonym(s):

Lignoceric acid, Tetracosanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)22COOH
CAS Number:
Molecular Weight:
368.64
Beilstein/REAXYS Number:
1728237
MDL number:

grade

analytical standard

assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

mp

82-86 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C24H48O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26/h2-23H2,1H3,(H,25,26)

InChI key

QZZGJDVWLFXDLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service