Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41976-U

Supelco

Ozone precursor / PAMS mix

57 components, 1 ppm each component in nitrogen, cylinder of 110 L (pi-marked cylinder), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

feature

standard type calibration

packaging

cylinder of 110 L (pi-marked cylinder)

manufacturer/tradename

Air Liquide America Specialty Gases LLC TAZ00007-110

concentration

1 ppm each component in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

General description

Ozone Precursor / PAMS Mix [57 components, 1 ppm each component in nitrogen, cylinder of 110 L (pi-marked cylinder)] manufactured by Air Liquide America Speciality Gases Toxic Organic (TO) is an excellent choice of calibration gas mixtures for use in monitoring ppm levels of toxic volatiles and semi-volatiles in air in accordance with U.S. Environmental Protection Agency′s TO-15/17 and Photochemical Assessment Monitoring Systems (PAMS) methodologies, and other related government regulations. The gas mixture is produced individually and gravimetrically using NIST traceable weights.

application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

The Ozone Precursor/PAMS (57 components) mix is supplied in an Extra-Life transportable aluminum cylinder which features Air Liquide′s exclusive Aculife cylinder treatment process for guaranteed mixture stability.

Blend tolerance and analytical accuracy = +/- 10%.

Specifications

Cylinder specifications:
Pressure:1800 psig
Outlet fitting:CGA 180
Weight:2.2lbs. (1kg)
Dimensions:3.25in D X 11.625in H (82.6mm X 295.3mm)

U.S. D.O.T. specs: 3AL2216

Analyte

Description

    Acetylene
    Benzene
    Butane
    1-Butene
    cis-2-Butene
    trans-2-Butene
    Cyclohexane
    Cyclopentane
    Decane
    1,3-Diethylbenzene
    1,4-Diethylbenzene
    2,2-Dimethylbutane
    2,3-Dimethylbutane
    2,3-Dimethylpentane
    2,4-Dimethylpentane
    Dodecane
    Ethane
    Ethylbenzene
    Ethylene
    3-Ethyltoluene
    2-Ethyltoluene
    4-Ethyltoluene
    Heptane
    Hexane
    1-Hexene
    2-Methylpropane
    2-Methylbutane
    Isoprene
    Cumene
    Methylcyclohexane
    Methylcyclopentane
    2-Methylheptane
    3-Methylheptane
    2-Methylhexane
    3-Methylhexane
    2-Methylpentane
    3-Methylpentane
    Nonane
    Octane
    Pentane
    1-Pentene
    cis-2-Pentene
    trans-2-Pentene
    Propane
    Propylbenzene
    Propylene
    Styrene
    Toluene
    1,2,3-Trimethylbenzene
    1,2,4-Trimethylbenzene
    Mesitylene
    2,2,4-Trimethylpentane
    2,3,4-Trimethylpentane
    Undecane
    Xylenes
    o-Xylene
    m/p-xylene
    See All (57)

pictograms

Gas cylinder

signalword

Warning

hcodes

Hazard Classifications

Press. Gas Compr. Gas

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service