Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41913

Supelco

EPA TO-14A GC-MS Tuning Standard

2 ppm bromofluorobenzene in nitrogen, pkg of 110 L, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116105

grade

analytical standard

agency

EPA TO-14

form

gas

packaging

pkg of 110 L

manufacturer/tradename

Scott Gas T1905-110

concentration

2 ppm bromofluorobenzene in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

General description

EPA TO-14A GC-MS Tuning Standard [2 ppm bromofluorobenzene in nitrogen, pkg of 110 L] may be used as a chemical reference standard for the qualitative calibration of air monitoring and industrial hygiene equipment. These standards are available in both liquid solutions and gaseous blends, to accommodate calibration of today′s variety of air monitoring and analysis devices and applications. Each standard includes a Certificate of Analysis summarizing the testing data. Each standard solution is provided in a flame-sealed amber ampoule. Each gaseous standard is provided in a transportable cylinder for ease of use in either the laboratory or the field.
These high pressure, transportable gas calibration blends for US EPA Method TO-14 ensure that laboratories can perform Method TO-14 and Clean Air Act analyses at a fraction of their former costs. Scott TO-14 gas blends are packaged in SCOTTY HP high pressure, transportable, laboratory-size cylinders. Scott TO-14 gas calibration blends are traceable to NIST reference mixtures (NIST SRM 1804), and are certified by Scott Gases for superior stability and accuracy.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Gas cylinder

signalword

Warning

hcodes

Hazard Classifications

Press. Gas Compr. Gas

Storage Class

2A - Gases

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service