Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41735

Supelco

Diol-functionalized silica gel spherical

75-200 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
23201100
NACRES:
SB.52

grade

standard

form

powder

composition

carbon content, 4.0-7.0%

technique(s)

LPLC: suitable

surface area

~300 m2/g

matrix

Silica

matrix active group

diol-functionalized silica

particle size

75-200 μm

pore size

110 Å pore size

separation technique

hydrophilic interaction (HILIC)

Looking for similar products? Visit Product Comparison Guide

General description

Diol-functionalized silica gel is less polar and has lower retention time when compared to normal phase silica gel.[1]

Application

Diol-functionalized silica gel may be used as stationary phase for liquid chromatographic analysis.

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

STOT SE 2 Inhalation

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G K Jayaprakasha et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 937, 25-32 (2013-09-10)
Rapid separation, characterization and quantitation of curcuminoids are important owing to their numerous pharmacological properties including antimicrobial, antiviral, antifungal, anticancer, and anti-inflammatory activities. In the present study, pseudo two-dimensional liquid flash chromatography was used for the separation of four curcuminoids

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service