Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41109

Supelco

1,3-Dichloropropane solution

5000 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein/REAXYS Number:
505960
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

ClCCCCl

InChI

1S/C3H6Cl2/c4-2-1-3-5/h1-3H2

InChI key

YHRUOJUYPBUZOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Rodriguez-Arnaiz
Environmental and molecular mutagenesis, 31(4), 390-401 (1998-07-08)
Biotransformation of several structurally related 2B compounds to reactive metabolites was evaluated in the somatic w/w+ assay of Drosophila melanogaster. Chemicals tested were the dichlorinated alkanes dichloromethane (DCM), 1,2-dichloroethane (DCE), and 1,3-dichloropropane (DCP); the thiouracil derivatives 5-methyl, 2-thiouracil (5M2TU), 6-methyl
S Shin et al.
Chemico-biological interactions, 97(3), 229-238 (1995-08-18)
Treatment of a lysosome-rich fraction from liver with 2-chloroethylethyl sulfide resulted in a dose-dependent release of arylsulfatase. The inclusion of Ca2+ enhanced the enzyme release by approximately 2.3-fold. The enhancing effect of Ca2+, showing an EC50 value of 30 mM
Daniel J Duffy et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(3), 659-671 (2004-01-30)
Ab initio calculations are reported for three of four possible conformers of 1,3-dichloropropane. The fourth conformer, with Cs symmetry, has a predicted enthalpy difference of more than 1500 cm(-1) from the most stable conformer from each calculation regardless of the
D W Herr et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 35(1), 31-48 (1997-01-01)
Previous research showed that acute exposure to dichloromethane (DCM) produced a selective reduction in peak N30 of flash evoked potentials (FEPs) in rats. In contrast, acute exposures to p-xylene or toluene selectively reduced FEP peak N160. The present experiments compared
Biological effect monitoring.
C T Evelo et al.
Archives of toxicology. Supplement. = Archiv fur Toxikologie. Supplement, 15, 268-277 (1992-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service