Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40572

Supelco

Methyl parathion solution

certified reference material, 1000 μg/mL in acetone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C8H10NO5PS/c1-12-15(16,13-2)14-8-5-3-7(4-6-8)9(10)11/h3-6H,1-2H3

InChI key

RLBIQVVOMOPOHC-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

1.4 °F - closed cup

flash_point_c

-17 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camila Bitencourt Mendes et al.
Environmental technology, 33(1-3), 167-172 (2012-04-24)
The assessment of vermicompost (VC) as a low-cost and alternative adsorbent for the removal of the pesticide methylparathion (MP) from an aqueous medium has been investigated by batch and column experiments. Parameters related to MP adsorption, i.e. equilibrium time (61.5
Scott A Trammell et al.
Chemical communications (Cambridge, England), 48(34), 4121-4123 (2012-03-22)
Using a low power green laser, we have demonstrated a rate acceleration of ~2-fold for the hydrolysis of methyl parathion by irradiating the plasmon absorption band of Au nanoparticles capped with a Cu(bpy) catalyst.
Shanli Yang et al.
Colloids and surfaces. B, Biointerfaces, 96, 75-79 (2012-04-20)
This paper proposed a direct electrodeposition approach to synthesis of graphene-chitosan (GR-CS) composite onto glassy carbon electrode (GCE) under controlled potential. This direct electrodeposition approach for the construction of GR-based hybrid was environmentally friendly, which would not involve the chemical
Thiago M R Araújo et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 48(4), 251-259 (2013-02-05)
This work presents a study of the abiotic degradation of commercially available methyl parathion in aqueous solution at two different concentrations (88 mg/L and 200 μg/L). The effects of solar irradiation and the presence of humic acids were evaluated and
Anirut Ekkhunnatham et al.
World journal of microbiology & biotechnology, 28(4), 1739-1746 (2012-07-19)
Methyl parathion hydrolase (MPH) from a methyl parathion-degrading Burkholderia cepacia indigenous to Thailand was purified to apparent homogeneity by three steps of column chromatography using Resource S, Sephadex G100, and Octyl Sepharose 4FF columns. Its molecular mass was determined to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service