Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40330

Supelco

2,4-D solution

certified reference material, 5000 μg/mL in acetonitrile

Synonym(s):

2,4-Dichlorophenoxyacetic acid solution, 2,4-D

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in acetonitrile

application(s)

agriculture
environmental

format

single component solution

Storage temp.

2-8°C

InChI

1S/C8H6Cl2O3/c9-5-1-2-7(6(10)3-5)13-4-8(11)12/h1-3H,4H2,(H,11,12)

Inchi Key

OVSKIKFHRZPJSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Hou et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 35(5), 675-678 (2012-12-12)
To explore and improve the utilization efficiency of Gentiana macrophylla resources. The germination of Gentiona macrophylla seeds were used as explants to induce calluses; Cultured on various culture media and condition for callus induction and subculture by test; The content
Bastian Niedrée et al.
Journal of environmental radioactivity, 115, 168-174 (2012-09-15)
In a soil microcosm experiment, the influence of low-level (137)Cs and (90)Sr contamination on the degradation of (14)C-ring-labeled 2,4-dichlorophenoxyacetic acid (2,4-D) was studied. Two differently treated soils (one native soil and one soil sterilized and reinoculated with a biotic soil
Erick R Bandala et al.
Environmental technology, 34(1-4), 363-371 (2013-03-28)
Estimation of scaling-up parameters for a site restoration process using a surfactant-enhanced soil washing (SESW) process followed by the application of advanced oxidation processes (Fenton and photo-Fenton) was performed. For the SESW, different parameters were varied and the soil washing
Jin-Liang Jia et al.
Nanotechnology, 23(49), 495710-495710 (2012-11-15)
Nearly monodisperse Au nanorods (NRs) with different aspect ratios were separated from home-synthesized polydisperse samples using a gradient centrifugation method. The morphology, size and its distribution, and photo-absorption property were analyzed by transmission electron microscopy, atomic force microscopy and UV-visible
Hong Sun et al.
Journal of applied toxicology : JAT, 32(8), 635-641 (2012-08-23)
The present study intended to develop receptor-mediated luciferase reporter gene assays to evaluate and compare the estrogen receptor (ER), androgen receptor (AR) and thyroid hormone receptor (TR) activities of target chemicals. Di-2-ethylhexyl-phthalate (DEHP), chlorpyrifos (CPF), 2,4-dichlorophenoxyacetic acid (2,4-D) and bisphenol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service