Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40323

Supelco

Hexachlorophene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

agency

EPA 8270

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C13H6Cl6O2/c14-6-2-8(16)12(20)4(10(6)18)1-5-11(19)7(15)3-9(17)13(5)21/h2-3,20-21H,1H2

InChI key

ACGUYXCXAPNIKK-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye-Jin Min et al.
Cancer letters, 276(2), 136-142 (2008-12-19)
Many studies have shown that the activation of beta-catenin signaling can promote oncogenesis, and it is therefore of interest to find agents that modulate this pathway. Recent work has shown using B lymphoma cells that infection by Epstein-Barr virus (EBV)
A Häntzschel et al.
Archives of toxicology, 73(10-11), 581-587 (2000-02-09)
N-methyl-D-aspartate (NMDA) and non-NMDA receptors were found to be involved in development of functional disorders caused by hexachlorophene. In order to specify the role of glutamate receptors we studied the protective effects of the selective antagonist of the kainate/(+/-)-alpha-amino-3-hydroxy-5-methylisoxazole-4-propionic acid
Yueming Zheng et al.
PloS one, 7(12), e51820-e51820 (2012-12-20)
The voltage-gated KCNQ1 potassium channel is expressed in cardiac tissues, and coassembly of KCNQ1 with an auxiliary KCNE1 subunit mediates a slowly activating current that accelerates the repolarization of action potential in cardiomyocytes. Mutations of KCNQ1 genes that result in
Joohee Jung et al.
Life sciences, 74(25), 3065-3074 (2004-04-15)
We examined the anti-estrogenic activity of 50 chemicals by the yeast two-hybrid assay and detected the activity of hexachlorophene, pentachlorophenol, and vitamin K3 (menadione), in that order. These chemicals were also observed to inhibit the transcriptional activity of 17beta-estradiol in
M H Wilcox et al.
The Journal of hospital infection, 56(2), 156-159 (2004-03-17)
We routinely phage-type Staphylococcus aureus isolates from high-risk inpatients each week. This surveillance approach previously identified a five-year outbreak of a methicillin-susceptible S. aureus strain (MSSA, PT 53,85), which affected 202 babies on a regional neonatal unit. We previously reported

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service