Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40251-U

Supelco

3-Methylphenol

certified reference material, 5000 μg/mL in methanol

Synonym(s):

m-Cresol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

agency

EPA TO-8
NIOSH 2001

feature

standard type calibration

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8H,1H3

InChI key

RLSSMJSEOOYNOY-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutsuko Koizumi et al.
The Journal of toxicological sciences, 28(2), 59-70 (2003-06-25)
To determine susceptibility of infants to 3-methylphenol, a repeated dose toxicity study was conducted with oral administration to newborn and young rats. In an 18-day newborn study from postnatal days 4 to 21 at doses of 30, 100 and 300
Claudio S Batista et al.
The Cochrane database of systematic reviews, (4)(4), CD006562-CD006562 (2010-04-16)
Genital warts are common and usually are harmless but can be painful and psychologically burdensome. Several local treatments can be used, including topical 5-Fluorouracil (5-FU). To determine the effectiveness and safety of 5-FU topical treatment for genital warts in nonimmunocompromised
K L Londry et al.
Canadian journal of microbiology, 45(6), 458-463 (1999-08-24)
The metabolism of cresols under sulfate-reducing conditions was investigated in Desulfotomaculum sp. strain Groll. This strain grows on a variety of aromatic compounds, including para- and meta- but not ortho-cresol. Degradation of p-cresol proceeded by oxidation reactions of the methyl
Qiang Chai et al.
Ying yong sheng tai xue bao = The journal of applied ecology, 17(9), 1624-1628 (2006-12-07)
With pot experiment, this paper studied the effects of water supply levels (45%, 60% and 75% of soil water hold capacity) and allelochemical 3-methylphenol (300 x 10(-6) mol x kg(-1) soil) on the soil microbial diversity and enzyme activity in
A J Simpson et al.
Journal of clinical pathology, 53(2), 159-160 (2000-04-18)
To re-examine the previously reported in vitro interaction of insulin with Burkholderia pseudomallei, in the light of a suggestion that the interaction may have resulted from the presence of the preservative m-cresol in commercial preparations. Broth culture studies of B

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service