Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40214

Supelco

1,3-Dichlorobenzene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
Beilstein/REAXYS Number:
956618
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1cccc(Cl)c1

InChI

1S/C6H4Cl2/c7-5-2-1-3-6(8)4-5/h1-4H

InChI key

ZPQOPVIELGIULI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J K Bibler-Muckelbauer et al.
Virology, 202(1), 360-369 (1994-07-01)
Crystallographic studies of human rhinovirus 14 (HRV14) crystals soaked with fragments of antiviral WIN compounds, at high concentrations (82-200 micrograms/ml), show the compounds bind into the hydrophobic beta-barrel (WIN pocket) of VP1. Two of these short compounds (5-[3,5-dimethyl-4-hydroxyphenyl]-2-methyltetrazole and phenol
Claudia Amorim et al.
Journal of hazardous materials, 211-212, 208-217 (2011-08-30)
Catalytic hydrodechlorination (HDC) is an effective means of detoxifying chlorinated waste. Involvement of spillover hydrogen is examined in gas phase dechlorination of chlorobenzene (CB) and 1,3-dichlorobenzene (1,3-DCB) over Pd and Ni. The catalytic action of single component Pd and Ni
D Mahadevan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 962-969 (2011-05-13)
The FT-IR and FT-Raman vibrational spectra of 1,3-dichlorobenzene (1,3-DCB) have been recorded using Bruker IFS 66 V Spectrometer in the range 4000-100 cm(-1). A detailed vibrational spectral analysis has been carried out and assignments of the observed fundamental bands have
N Yoshida et al.
Analytical chemistry, 73(18), 4417-4421 (2001-09-29)
A distributed-feedback dye laser has been developed for achieving the efficient multiphoton ionization of chlorobenzene and dichlorobenzene, that is, precursor molecules of dioxins. This tunable picosecond laser with a narrow spectral line width, that is, a nearly transform-limited pulse, provides
T Umemura et al.
Toxicology and applied pharmacology, 137(2), 268-274 (1996-04-01)
The acute hepatotoxicity of isomers of dichlorobenzene (o-, m-, and p-DCB) was compared in livers of male B6C3F1 mice at different time points after a single intragastric administration. The highest doses of o-, m-, and p-DCB administered, 300, 300, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service