Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40176-U

Supelco

HC 8270 LCS Spike Mix

certified reference material, ampule of 25 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

packaging

ampule of 25 mL

technique(s)

gas chromatography (GC): suitable

format

multi-component solution

storage temp.

−20°C

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acenaphthene
    Acenaphthylene
    Aniline
    Anthracene
    Azobenzene
    Benz[a]anthracene
    Benzo[b]fluoranthene
    Benzo[k]fluoranthene
    Benzo[ghi]perylene
    Benzo[a]pyrene
    Benzyl alcohol
    Benzyl butyl phthalate
    Bis(2-chloroethyl) ether
    Bis-(2-chloroisopropyl) ether
    Bis(2-chloroethoxy)methane
    bis-2-ethylhexyl adipate
    Bis(2-ethylhexyl) phthalate
    4-Bromodiphenyl ether
    Carbazole
    4-Chloroaniline
    4-Chlorodiphenyl ether
    4-Chloro-3-methylphenol
    2-Chlorophenol
    2-Chloronaphthalene
    m-Cresol
    o-Cresol
    p-Cresol
    Dibenz[a,h]anthracene
    Dibenzofuran
    Di-n-butyl phthalate
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    Dichloromethane
    2,4-Dichlorophenol
    2,4-Dimethylphenol
    Dimethyl phthalate
    1,2-Dinitrobenzene
    1,3-Dinitrobenzene
    1,4-Dinitrobenzene
    2,4-Dinitrophenol
    2,4-Dinitrotoluene
    2,6-Dinitrotoluene
    Dioctyl phthalate
    Fluoranthene
    Fluorene
    Hexachlorobenzene
    Hexachloro-1,3-butadiene
    Hexachlorocyclopentadiene
    Hexachloroethane
    Indeno[1,2,3-cd]pyrene
    Isophorone
    2-Methyl-4,6-dinitrophenol
    1-Methylnaphthalene
    2-Methylnaphthalene
    Naphthalene
    2-Nitroaniline
    3-Nitroaniline
    4-Nitroaniline
    Nitrobenzene
    N-Nitrosodimethylamine
    N-Nitrosodiphenylamine
    N-Nitrosodi-n-propylamine
    2-Nitrophenol
    4-Nitrophenol
    Pentachlorophenol
    Phenanthrene
    Phenol
    Pyrene
    Pyridine
    2,3,4,6-Tetrachlorophenol
    2,3,5,6-Tetrachlorophenol
    1,2,4-Trichlorobenzene
    2,4,5-Trichlorophenol
    2,4,6-Trichlorophenol
    See All (75)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Repr. 2 - STOT RE 2 - STOT SE 1

target_organs

Blood, Eyes,Central nervous system

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service