40104-U
Methanolic HCl
3 M HCl in methanol (3N), pkg of 1 × 2 L
Synonym(s):
Methanolic HCl, Hydrochloric acid solution
About This Item
Recommended Products
grade
derivatization grade ((GC derivatization))
form
liquid
reaction suitability
reagent type: derivatization reagent
reaction type: Esterifications
packaging
pkg of 1 × 2 L
concentration
3 M HCl in methanol (3N)
density
0.79 g/cm3
storage temp.
−20°C
InChI
1S/ClH/h1H
InChI key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
General description
Features and Benefits
- Derivatization of fatty acids, particularly volatile (short chain) fatty acids.
- Clean reaction (no side reaction) with volatile by-products.
- Provides clean, fast, quantitative derivatization.
related product
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3
target_organs
Eyes,Central nervous system, Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
39.9 °F
flash_point_c
4.4 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service