Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40099

Supelco

Heptachlor expoxide isomer B solution

certified reference material, 1000 μg/mL in methanol

Synonym(s):

Heptachlor exo-epoxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5Cl7O
CAS Number:
Molecular Weight:
389.32
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

[H][C@]12O[C@@]1([H])[C@@]3([H])[C@@]([H])([C@H]2Cl)C4(Cl)C(Cl)=C(Cl)C3(Cl)C4(Cl)Cl

InChI

1S/C10H5Cl7O/c11-3-1-2(4-5(3)18-4)9(15)7(13)6(12)8(1,14)10(9,16)17/h1-5H/t1-,2+,3+,4-,5+,8?,9?/m0/s1

InChI key

ZXFXBSWRVIQKOD-TTWWGNBVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - Solvent

flash_point_c

9.7 °C - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guadalupe Prado et al.
Mutation research, 673(2), 87-91 (2009-01-17)
The genotoxic potential of the organochlorine insecticides heptachlor (HC) and its metabolite heptachlor epoxide (HCE) has been evaluated in TK6 cells, a well-established human lymphoblastoid cell line. Genotoxicity has been determined by scoring the induction of DNA breaks in the
Chhatra Mani Sharma et al.
Environmental pollution (Barking, Essex : 1987), 157(8-9), 2452-2458 (2009-03-31)
Organochlorine pollutants in the major fish species (pike Esox lucius, perch Perca fluviatilis, and roach Rutilus rutilus) of Lake Arungen, Norway, were investigated after an extensive removal of large pike in 2004. The organochlorine pollutants detected in fish liver samples
M E Hansen et al.
Toxicology, 160(1-3), 139-153 (2001-03-14)
The effects of the organochlorine (OC) liver tumor promoter heptachlor epoxide (HE; 0, 0.1, 1, 10, and 50 microM) on several cellular tumor promoter-sensitive parameters were studied in mouse 1c1c7 hepatoma cells in an effort to identify the most sensitive
Ulrike Luderer et al.
Environmental research, 121, 84-94 (2012-12-01)
For 15 months in 1981-1982, the commercial milk supply on the Hawaiian island of Oahu was contaminated with heptachlor epoxide, a metabolite of the insecticide heptachlor, resulting in gestational and/or lactational exposure to offspring of women who drank cow milk
Wim J M Hegeman et al.
Reviews of environmental contamination and toxicology, 173, 85-116 (2002-01-05)
Enantiomer fractions (EFs) of chiral compounds have been used to explain the mechanisms of enantiomer enrichment in air, soil, water, and biota. The EFs were calculated from enantiomeric ratios (ERs) of chiral compounds measured by researchers during the past 10

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service