Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40059

Supelco

N-Nitrosodimethylamine (NDMA) solution

5000 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105

grade

analytical standard

agency

EPA TO-7

feature

standard type calibration

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C2H6N2O/c1-4(2)3-5/h1-2H3

InChI key

UMFJAHHVKNCGLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

András P Keszei et al.
The American journal of clinical nutrition, 97(1), 135-146 (2012-11-30)
Dietary N-nitroso compounds and endogenous nitrosation are important carcinogenic factors, but human evidence of their role is scarce for esophageal cancer and inconsistent for gastric cancer. We studied the relation between risks of esophageal and gastric cancer subtypes and dietary
Ying Han et al.
Water research, 47(1), 216-224 (2012-10-23)
N-Nitrosodimethylamine (NDMA) is known as the disinfection by-product and the pollutant in the source water. Reduction with zero-valent zinc (Zn(0)) was investigated as a potential technology to treat NDMA. The results showed that Zn(0) was effective for NDMA reduction at
J W Hatt et al.
Chemosphere, 91(1), 83-87 (2012-12-06)
Concern over prospective levels of N-nitrosodimethylamine (NDMA) in waters has increased in recent years due to its disinfection byproduct formation potential from chloramination. It has been mooted that this is promoted by organic precursors from municipal wastewaters, such that there
David Hanigan et al.
Environmental science & technology, 46(22), 12630-12639 (2012-10-31)
Activated carbon (AC) has been shown to remove precursors of halogenated disinfection byproducts. Granular and powdered activated carbon (GAC, PAC) were investigated for their potential to adsorb N-nitrosodimethylamine (NDMA) precursors from blends of river water and effluent from a wastewater
Man-Chun Huang et al.
Food chemistry, 138(1), 227-233 (2012-12-26)
A sensitive procedure, microwave-assisted extraction (MAE) coupled dispersive micro solid-phase extraction (D-μ-SPE), was developed to extract N-nitrosodimethylamine (NDMA) and other six volatile N-nitrosamines (NAms) from meat products. Parameters affecting the efficiency of MAE and D-μ-SPE were systematically investigated. For MAE

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service