Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40048-U

Supelco

Antioxidant Kit 2, Phenolics

(individually packaged in quantities indicated), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

kit of 1

concentration

(individually packaged in quantities indicated)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-30°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    PROPYL GALLATE - PHR1118 1000MG
    TERT BUTYLHYDROQUINONE (TBHQ) - PHR3512 500MG
    NORDIHYDROGUAIARETIC ACID - PHR3509 500MG
    BUTYLATED HYDROXYANISOLE - PHR1304 1000MG
    BUTYLATED HYDROXYTOLUENE - PHR1117 1000MG
    3,5 DI-TERT-BUTYL-4-HYDROXYBENZYL ALCOHOL - PHR3507 500MG
    LAURYL GALLATE - PHR3510 500MG
    OCTYL GALLATE - PHR3511 500MG
    ETHYOXYQUIN - PHR3513 500MG
    See All (9)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service