Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40046

Supelco

Bromodichloromethane solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHBrCl2
CAS Number:
Molecular Weight:
163.83
EC Number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

agency

EPA 601

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/CHBrCl2/c2-1(3)4/h1H

InChI key

FMWLUWPQPKEARP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S R Bielmeier et al.
Toxicology in vitro : an international journal published in association with BIBRA, 21(5), 919-928 (2007-03-09)
Bromodichloromethane (BDCM), a drinking water disinfection by-product, causes pregnancy loss, i.e. full-litter resorption, in F344 rats when treated during the luteinizing hormone (LH)-dependent period. This effect is associated with reduced maternal serum progesterone (P) and LH levels, suggesting that BDCM
Richard J Summerhayes et al.
Epidemiology (Cambridge, Mass.), 23(1), 15-22 (2011-12-14)
Trihalomethanes in drinking water have been associated with higher occurrence of small-for-gestational-age (SGA) births, although results have been inconsistent. We geocoded residential address for mother of live, singleton, term births to 33 water distribution systems in a large metropolitan area
National Toxicology Program technical report series, (532)(532), 1-248 (2006-06-03)
Bromodichloromethane is a by-product of the chlorination of drinking water. It is formed by the halogen substitution and oxidation reactions of chlorine with naturally occurring organic matter (e.g., humic or fulvic acids) in water containing bromide. Bromodichloromethane has been shown
Minh T Do et al.
Environmental health perspectives, 113(4), 418-424 (2005-04-07)
Chlorination disinfection by-products (CDBPs) are produced during the treatment of water with chlorine to remove bacterial contamination. CDBPs have been associated with an increased risk of bladder cancer. There is also some evidence that they may increase the risk of
Suvarthi Das et al.
Toxicology and applied pharmacology, 269(3), 297-306 (2013-02-27)
Today's developed world faces a major public health challenge in the rise in the obese population and the increased incidence in fatty liver disease. There is a strong association among diet induced obesity, fatty liver disease and development of nonalcoholic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service