Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40019

Supelco

2,4,6-Trichlorophenol solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

agency

EPA 552

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C6H3Cl3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H

InChI key

LINPIYWFGCPVIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Yan et al.
Huan jing ke xue= Huanjing kexue, 32(10), 3059-3066 (2012-01-28)
Photolytic circulating-bed biofilm reactor (PCBBR) and internal loop photolytic-biological reactor (ILPBR) were respectively used for degradation of phenol, 2, 4, 6-trichlorophenol (TCP) and sulfamethoxazole (SMX). Experimental results indicated that the rates of phenol, TCP and SMX removal by coupled photolysis
James Huff
Chemosphere, 89(5), 521-525 (2012-07-04)
Carcinogenesis bioassays were conducted by giving 2,4,6-trichlorophenol [2,4,6-TCP] in feed to groups of 50 male and female Fischer rats and male B6C3F1 mice for two years. Dietary concentrations were 0 [20/group], 5000 [0.5%], or 10,000 [1%] ppm. Female mice began
Supreet S Bahga et al.
Electrophoresis, 33(6), 1048-1059 (2012-04-25)
We present a novel method of creating concentration cascade of leading electrolyte (LE) in isotachophoresis (ITP) by using bidirectional ITP. ITP establishes ion-concentration shock waves between high-mobility LE and low-mobility trailing electrolyte (TE) ions. In bidirectional ITP, we set up
Minhee Kim et al.
Chemosphere, 89(3), 262-268 (2012-05-17)
The well-known cosolvency-induced sorption model is not applicable to predict the sorption of carboxylic acids in cosolvent system. To investigate the phenomenon, sorption and solubility of chlorinated phenols (2,4-dichlorophenol (2,4-DCP) and 2,4,6-trichlorophenol (2,4,6-TCP)) and carboxylic acids (benzoic acid and 2,4-dichlorophenoxyacetic
Stefan Franzen et al.
The journal of physical chemistry. B, 116(5), 1666-1676 (2011-12-14)
Kinetic and structural studies have shown that peroxidases are capable of the oxidation of 2,4,6-trichlorophenol (2,4,6-TCP) to 2,6-dichloro-1,4-benzoquinone (2,6-DCQ). Further reactions of 2,6-DCQ in the presence of H(2)O(2) and OH(-) yield 2,6-dichloro-3-hydroxy-1,4-benzoquinone (2,6-DCQOH). The reactions of 2,6-DCQ have been monitored

Protocols

Analytical standard separation of various phenols for research and industrial applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service