Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38703

Supelco

Rotenone

reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22O6
CAS Number:
Molecular Weight:
394.42
Beilstein/REAXYS Number:
6773081
MDL number:
UNSPSC Code:
41116107

grade

reference material

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

bp

210-220 °C/0.5 mmHg (lit.)

mp

159-164 °C (lit.)

format

neat

Storage temp.

2-8°C

SMILES string

COc1cc2OCC3Oc4c5C[C@@H](Oc5ccc4C(=O)C3c2cc1OC)C(C)=C

InChI

1S/C23H22O6/c1-11(2)16-8-14-15(28-16)6-5-12-22(24)21-13-7-18(25-3)19(26-4)9-17(13)27-10-20(21)29-23(12)14/h5-7,9,16,20-21H,1,8,10H2,2-4H3/t16-,20-,21+/m1/s1

Inchi Key

JUVIOZPCNVVQFO-HBGVWJBISA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Biochem/physiol Actions

Inhibitor of mitochondrial electron transport.
Rotenone is an inhibitor of mitochondrial electron transport at nicotinamide adenine dinucleotide (NADH):ubiquinone oxidoreductase. It is readily absorbed through the exoskeletons of arthropods, but poorly absorbed cutaneously or from the gastrointestinal tract of mammals. Rotenone acts as a neurotoxic agent which can produce Parkinson-like condition to serve as an animal model for the study of etiology and interventions. The primary mechanism of action of rotenone is complex I inhibition, followed by the generation of oxidative stress and oxidative damage. Because rotenone is lipophilic, it readily penetrates the brain, where it attaches to and inhibits mitochondrial complex I of electron chain transport (ETC).

Pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service