Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33355

Supelco

Methanolic HCl

3 M HCl in methanol (3N), derivatization grade (GC derivatization), ampule of 20 × 1 mL

Synonym(s):

Hydrochloric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
UNSPSC Code:
41116105
PubChem Substance ID:
Product 33355 is not currently sold in your country. Contact Technical Service

grade

derivatization grade (GC derivatization)

assay

99.9% (purity)

form

liquid

reaction suitability

reagent type: derivatization reagent
reaction type: Esterifications

packaging

ampule of 20 × 1 mL

concentration

3 M HCl in methanol (3N)

storage temp.

2-8°C

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanolic HCl (hydrochloric acid in methanol) is particularly useful for preparing methyl esters of volatile (short chain) fatty acids. Fatty acids are esterified by heating them with an anhydrous alcohol (e.g., methanol) in the presence of an acidic catalyst (e.g., HCl) in a sealed vessel at a high temperature for a short time. In the reaction, a fatty acid molecule and an alcohol molecule are joined, with the release of a water molecule. The derivatives can be quickly and easily recovered, quantitatively, from the reaction medium.

Features and Benefits

  • Derivatization of fatty acids, particularly volatile (short chain) fatty acids.

  • Clean reaction (no side reaction) with volatile by-products.

  • Provides clean, fast, quantitative derivatization.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Met. Corr. 1 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of limiting polyunsaturated fatty acids in Daphnia galeata using a new method to enrich food algae with single fatty acids.
Von EE.
Limnology and Oceanography, 47 (6), 1764-1773 (2002)
Mitochondrial Function, Part B: Mitochondrial Protein Kinases, Protein Phosphatases and Mitochondrial Diseases.
William SA and Murphy AN.
Science, 88-89 (2009)
Nicolas Cetraro et al.
The Analyst, 144(19), 5848-5855 (2019-09-05)
The carbon-carbon double bond positions of unsaturated fatty acids can have markedly different effects on biological function and also serve as biomarkers of disease pathology, dietary history, and species identity. As such, there is great interest in developing methods for
Practical Gas Chromatography: A Comprehensive Reference.
Wilder KD and Engewald W.
Science, 613-614 (2014)
Enamel color modifications by controlled hydrochloric acid pumice abrasion: a review with case summaries.
T C Adams
Journal (Indiana Dental Association), 66(5), 23-26 (1987-09-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service