Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33151

Supelco

BSA+TMCS+TMSI

3:2:3, pkg of 144 × 0.1 mL

Synonym(s):

Chlorotrimethylsilane, N,O-Bis(trimethylsilyl)acetamide, TSIM, Trimethylchlorosilane, Trimethylsilyl chloride, Trimethylsilylimidazole

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

description

3:2:3 (SYLON BTZ)

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 144 × 0.1 mL

concentration

(3:2:3)

technique(s)

GC/MS: suitable (SIM)

Looking for similar products? Visit Product Comparison Guide

General description

Reagent mixture consisting of BSA+TMCS+TMSI when heated with pyridine and steroid at 60°C for 12 hours, converts 11- and the tertiary 17-hydroxyl groups to trimethylsilyl ethers.[1]

application

It was used as to perform derivatization of the sample for multi-mycotoxin analysis using gas chromatography (GC) coupled to triple quadrupole instrument (QqQ).[2] It was also used as derivatizing reagent in the determination of trichothecenes, patulin and zearalenone using GC-MS/MS.[3]

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

28.4 °F - closed cup

flash_point_c

-2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Casparrini et al.
Chemistry and Physics of Lipids, 3, 1-1 (1969)
J. Throck Watson, O. David Sparkman
Introduction to Mass Spectrometry: Instrumentation, Applications, and Strategies for Data Interpretation, 588-588 (2007)
B W Wilson et al.
Agents and actions, 6(1-3), 176-179 (1976-02-01)
A GC-MS assay for PGE2 and PGF2alpha has been described. The method is highly specific and sensitive down to the sub-picogram level. The assay has been used to measure the time dependence of PG concentration in carrageenan-induced inflammatory exudates and
Yelko Rodríguez-Carrasco et al.
Food chemistry, 146, 212-219 (2013-11-02)
An analytical protocol based on QuEChERS and gas chromatography-tandem mass spectrometry (GC-MS/MS) was successfully applied for the determination of trichothecenes, patulin and zearalenone in 182 milled grain-based samples. The analytical method was validated following the SANCO 1495/2011 document. LOQs were
A gas chromatography-mass spectrometry method for the quantitative analysis of melatonin in plasma and cerebrospinal fluid.
B W Wilson et al.
Analytical biochemistry, 81(2), 283-291 (1977-08-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service