Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33148

Supelco

BSTFA + TMCS, 99:1

pkg of 20 × 1 mL

Synonym(s):

Chlorotrimethylsilane, N,O-Bis(trimethylsilyl)trifluoroacetamide, Trimethylchlorosilane, Trimethylsilyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

description

(SYLON BFT)

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 20 × 1 mL

concentration

(BSTFA+TMCS, 99:1)

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for the derivatization of alkylphosphonic acids, aminoalkylphosphonic acids, glycerophospholipids, polyphosphoinositides, steroid phosphates, tyrosine, tryptophan and metabolites.

Other Notes

Reagent for isothiocyanate (amino compounds), oxime, trimethylsilyl, trimethylsilyl deacylated polyphosphoinositides and trimethylsilyl glycerophosphate esters.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

75.2 °F - closed cup

flash_point_c

24 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.R. Knapp
Handbook of Analytical Derivatization Reactions, 214-215 (1979)
T.J. Cicero and W.R. Sherman
Biochemical and Biophysical Research Communications, 42, 728-728 (1971)
P.W. Albro and L. Fishbein
Journal of Chromatography A, 55, 297-297 (1971)
Mass spectral analysis glycerophospholipids.
J H Duncan et al.
Biochemistry, 10(6), 927-932 (1971-03-16)
D.J. Harvey and M.G. Horning
Journal of Chromatography A, 79, 65-65 (1973)

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service